CAS RN 516-42-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-Cortolone, TRC
CAS: 516-42-7 Molecular Formula: C21 H34 O5 Molecular Weight (g/mol): 366.49 Synonym: 3α,17,20α,21-Tetrahydroxy-5β-pregnan-11-one,20α-Cortolone,3α,17,20α,21-Tetrahydroxy-5β-pregnan-11-one,3α,17α,20α,21-Tetrahydroxy-5β-pregnan-11-one,5β-Pregnan-3α,17α,20α,21-tetrol-11-one,5β-Pregnane-3α,17,20α,21-tetrol-11-one,5β-Pregnane-3α,17α,20α,21-tetrol-11-one,Cortolon,Cortolone,NSC 59872 IUPAC Name: (3R,5R,8S,9S,10S,13S,14S,17R)-17-[(1S)-1,2-dihydroxyethyl]-3,17-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-11-one SMILES: C[C@]12CC[C@@H](O)C[C@H]1CC[C@H]3[C@@H]4CC[C@](O)([C@@H](O)CO)[C@@]4(C)CC(=O)[C@H]23