accessibility menu, dialog, popup

UserName

CAS RN 51676-74-5

IUPAC Name: 1-chloro-5-methyl-2,4-dinitrobenzene
Synonyms: 1-chloro-5-methyl-2,4-dinitrobenzene
Molecular Weight (g/mol): 216.58
Molecular Formula: C7H5ClN2O4
InChi Key: KPDPGZNHKMJEFZ-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results