accessibility menu, dialog, popup

UserName

CAS RN 51694-22-5

ONOSeN

CAS RN 51694-22-5

IUPAC Name: [(2-nitrophenyl)selanyl]formonitrile
Synonyms: [(2-nitrophenyl)selanyl]formonitrile
Molecular Weight (g/mol): 227.09
Molecular Formula: C7H4N2O2Se
InChi Key: LHBLJWULWKQRON-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1[Se]C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)