accessibility menu, dialog, popup

UserName

HOOOCl

CAS RN 5197-62-6

IUPAC Name: 2-[2-(2-chloroethoxy)ethoxy]ethan-1-ol
Synonyms: 2-[2-(2-chloroethoxy)ethoxy]ethanol
Molecular Weight (g/mol): 168.62
Molecular Formula: C6H13ClO3
InChi Key: KECMLGZOQMJIBM-UHFFFAOYSA-N
SMILES: OCCOCCOCCCl
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)