accessibility menu, dialog, popup

UserName

CAS RN 52-89-1

IUPAC Name: hydrogen (2R)-2-amino-3-sulfanylpropanoic acid chloride
Synonyms: cysteine hcl
Molecular Weight (g/mol): 157.61
Molecular Formula: C3H8ClNO2S
InChi Key: IFQSXNOEEPCSLW-DKWTVANSSA-N
SMILES: [H+].[Cl-].N[C@@H](CS)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (5)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)