accessibility menu, dialog, popup

UserName

CAS RN 521074-81-7

IUPAC Name: N-ethyl-2-(piperidin-4-yloxy)acetamide
Synonyms: N-ethyl-2-(piperidin-4-yloxy)acetamide
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: FICKGLKQSSSVOL-UHFFFAOYSA-N
SMILES: CCNC(=O)COC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results