CAS RN 5217-47-0
CAS RN 5217-47-0
IUPAC Name:
1,3-diethyl-2-sulfanylidene-1,3-diazinane-4,6-dione
Synonyms:
1,3-diethyl-2-sulfanylidene-1,3-diazinane-4,6-dione
Molecular Weight (g/mol):
200.26
Molecular Formula:
C8H12N2O2S
InChi Key:
SHBTUGJAKBRBBJ-UHFFFAOYSA-N
SMILES:
CCN1C(=O)CC(=O)N(CC)C1=S
No Refinements
1
–
0
of
0
results
1
1,3-Diethyl-2-thiobarbituric acid, 99%
CAS: 5217-47-0 Molecular Formula: C8H12N2O2S Molecular Weight (g/mol): 200.26 MDL Number: MFCD00006676 InChI Key: SHBTUGJAKBRBBJ-UHFFFAOYSA-N Synonym: 1,3-diethyl-2-thiobarbituric acid,1,3-diethylthiobarbituric acid,n,n'-diethylthiobarbituric acid,n,n'-diethyl-2-thiobarbituric acid,1,3-diethyl-2-thioxodihydropyrimidine-4,6 1h,5h-dione,unii-bxb40m6a7w,4,6 1h,5h-pyrimidinedione, 1,3-diethyldihydro-2-thioxo,barbituric acid, 1,3-diethyl-2-thio,bxb40m6a7w,1,3-diethyldihydro-2-thioxopyrimidine-4,6 1h,5h-dione PubChem CID: 78890 IUPAC Name: 1,3-diethyl-2-sulfanylidene-1,3-diazinane-4,6-dione SMILES: CCN1C(=O)CC(=O)N(CC)C1=S