CAS RN 523-39-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Menaquinone 9, TRC
CAS: 523-39-7 Molecular Formula: C55 H78 O2 Molecular Weight (g/mol): 771.21 Synonym: 2-Methyl-3-[(2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-nonamethyl-2,6,10,14,18,22,26,30,34-hexatriacontanonaen-1-yl]-1,4-naphthalenedione,Vitamin K2(45) (6CI),MK9,Menaquinone MK 9,Vitamin MK 9; IUPAC Name: 2-methyl-3-[(2E,6E,10E,14E,18E,21E,25E,29E)-3,7,11,15,19,22,26,30,34-nonamethylpentatriaconta-2,6,10,14,18,21,25,29,33-nonaenyl]naphthalene-1,4-dione SMILES: CC(=CCC\C(=C\CC\C(=C\CC\C(=C\C\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC1=C(C)C(=O)c2ccccc2C1=O)\C)\C)\C)\C)\C)\C)\C)\C)C