accessibility menu, dialog, popup

UserName

OO

CAS RN 525-82-6

IUPAC Name: 2-phenyl-4H-chromen-4-one
Synonyms: flavone cromarile flavone, 14c-labeled chromocor asmacoril cromaril 2-phenyl-4-chromone 2-phenyl-4-benzopyron flavone (van) (8CI) 14C-labeled flavone
Molecular Weight (g/mol): 222.24
Molecular Formula: C15H10O2
InChi Key: VHBFFQKBGNRLFZ-UHFFFAOYSA-N
SMILES: O=C1C=C(OC2=CC=CC=C12)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results