accessibility menu, dialog, popup

UserName

CAS RN 52548-00-2

CH3H3CBrF

CAS RN 52548-00-2

IUPAC Name: 1-bromo-4-fluoro-2,3-dimethylbenzene
Synonyms: 1-bromo-4-fluoro-2,3-dimethylbenzene
Molecular Weight (g/mol): 203.05
Molecular Formula: C8H8BrF
InChi Key: OYWLDZXWISLBMC-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(Br)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results