CAS RN 52554-28-6
CAS RN 52554-28-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2,3,5-Tri-O-acetyl-D-ribofuranoside, TRC
CAS: 52554-28-6 Molecular Formula: C12H18O8 Molecular Weight (g/mol): 290.27 IUPAC Name: [(2R,3R,4R)-3,4-diacetyloxy-5-methoxyoxolan-2-yl]methyl acetate SMILES: COC1O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C