CAS RN 526-37-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-Dihydroxy-4-methylbenzaldehyde, TRC
CAS: 526-37-4 Molecular Formula: C8 H8 O3 Molecular Weight (g/mol): 152.1473 Synonym: Atranol (6CI),γ-Resorcylaldehyde, 4-methyl- (7CI,8CI),2,6-Dihydroxy-4-methylbenzaldehyde,2,6-Dihydroxy-p-methylbenzaldehyde,4-Methyl-2,6-dihydroxybenzaldehyde,p-Orsellinaldehyde,Benzaldehyde, 2,6-dihydroxy-4-methyl- IUPAC Name: 2,6-dihydroxy-4-methylbenzaldehyde SMILES: Cc1cc(O)c(C=O)c(O)c1