accessibility menu, dialog, popup

UserName

CAS RN 5271-38-5

CH3SHO

CAS RN 5271-38-5

IUPAC Name: 2-(methylsulfanyl)ethan-1-ol
Synonyms: 2-(methylthio)ethanol
Molecular Weight (g/mol): 92.16
Molecular Formula: C3H8OS
InChi Key: WBBPRCNXBQTYLF-UHFFFAOYSA-N
SMILES: CSCCO
Boiling Point 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)