accessibility menu, dialog, popup

UserName

CAS RN 5279-32-3

IUPAC Name: 5,6-dibromo-2H-1,3-benzodioxole
Synonyms: 5,6-dibromo-2H-1,3-benzodioxole
Molecular Weight (g/mol): 279.92
Molecular Formula: C7H4Br2O2
InChi Key: WPYAICCSYGUFTK-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OCO2)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results