accessibility menu, dialog, popup

UserName

CAS RN 5292-43-3

CH3CH3H3COOBr

CAS RN 5292-43-3

IUPAC Name: tert-butyl 2-bromoacetate
Synonyms: tert-butyl 2-bromoacetate
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: BNWCETAHAJSBFG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CBr
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)