accessibility menu, dialog, popup

UserName

CAS RN 531-57-7

ClCH3HNNNH2S

CAS RN 531-57-7

IUPAC Name: 3-amino-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride
Synonyms: 3-amino-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride
Molecular Weight (g/mol): 277.77
Molecular Formula: C13H12ClN3S
InChi Key: OBKVLZKRUVYQNQ-UHFFFAOYSA-M
SMILES: [Cl-].CNC1=CC=C2N=C3C=CC(N)=CC3=[S+]C2=C1
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)