CAS RN 53112-33-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methyl-1,2,3,4-tetrahydroisoquinoline Hydrochloride, TRC
CAS: 53112-33-7 Molecular Formula: C10 H13 N . H Cl Molecular Weight (g/mol): 183.68 Synonym: Isoquinoline, 1,2,3,4-tetrahydro-2-methyl-, hydrochloride (1:1) (ACI),Isoquinoline, 1,2,3,4-tetrahydro-2-methyl-, hydrochloride (7CI, 9CI),2-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride,N-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride,1,2,3,4-Tetrahydro-2-methylisoquinoline hydrochloride IUPAC Name: 2-methyl-3,4-dihydro-1H-isoquinoline;hydrochloride SMILES: Cl.CN1CCc2ccccc2C1