accessibility menu, dialog, popup

UserName

AgOO

CAS RN 532-31-0

IUPAC Name: benzoic acid silver
Synonyms: silver benzoate
Molecular Weight (g/mol): 229.99
Molecular Formula: C7H6AgO2
InChi Key: GQCYXEOBZQXHPO-UHFFFAOYSA-N
SMILES: [Ag].OC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results