accessibility menu, dialog, popup

UserName

CH3H3CCH3OHNNBOH3CH3CH3CCH3O

CAS RN 532391-30-3

IUPAC Name: 2,2-dimethyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]propanamide
Synonyms: 2,2-dimethyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]propanamide
Molecular Weight (g/mol): 304.2
Molecular Formula: C16H25BN2O3
InChi Key: JTGWGXPOGSUYCT-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=NC=CC=C1B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results