accessibility menu, dialog, popup

UserName

HOOHOHOHO

CAS RN 533-87-9

IUPAC Name: 9,10,16-trihydroxyhexadecanoic acid
Synonyms: aleuritic acid
Molecular Weight (g/mol): 304.43
Molecular Formula: C16H32O5
InChi Key: MEHUJCGAYMDLEL-UHFFFAOYNA-N
SMILES: OCCCCCCC(O)C(O)CCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)