accessibility menu, dialog, popup

UserName

CAS RN 5330-98-3

ONOSCl

CAS RN 5330-98-3

IUPAC Name: 2-chloro-3-nitrothiophene
Synonyms: 2-chloro-3-nitrothiophene
Molecular Weight (g/mol): 163.58
Molecular Formula: C4H2ClNO2S
InChi Key: OFPRGOSJWUNETN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)SC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results