accessibility menu, dialog, popup

UserName

CAS RN 5331-92-0

OHNClCl(E)

CAS RN 5331-92-0

IUPAC Name: (E)-N-[(3,4-dichlorophenyl)methylidene]hydroxylamine
Synonyms: (E)-N-[(3,4-dichlorophenyl)methylidene]hydroxylamine
Molecular Weight (g/mol): 190.02
Molecular Formula: C7H5Cl2NO
InChi Key: ROBIUDOANJUDHD-ONNFQVAWSA-N
SMILES: O\N=C\C1=CC=C(Cl)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results