accessibility menu, dialog, popup

UserName

BrNOO

CAS RN 5332-26-3

IUPAC Name: 2-(bromomethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(bromomethyl)isoindole-1,3-dione
Molecular Weight (g/mol): 240.06
Molecular Formula: C9H6BrNO2
InChi Key: UUSLLECLCKTJQF-UHFFFAOYSA-N
SMILES: BrCN1C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results