accessibility menu, dialog, popup

UserName

NH2NN

CAS RN 5356-71-8

IUPAC Name: 1,3-diphenyl-1H-pyrazol-5-amine
Synonyms: 2,5-diphenylpyrazol-3-amine
Molecular Weight (g/mol): 235.29
Molecular Formula: C15H13N3
InChi Key: SXOFMEWDEKEVJU-UHFFFAOYSA-N
SMILES: NC1=CC(=NN1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)