CAS RN 53912-85-9
CAS RN 53912-85-9
IUPAC Name:
hydrogen 2-[(2S)-pyrrolidin-2-yl]acetic acid chloride
Synonyms:
hydrogen (2S)-pyrrolidin-2-ylacetic acid chloride
Molecular Weight (g/mol):
165.62
Molecular Formula:
C6H12ClNO2
InChi Key:
VQDACVOAOJQTPR-JEDNCBNOSA-N
SMILES:
[H+].[Cl-].OC(=O)C[C@@H]1CCCN1
1
–
1
of
1
results
1
L-beta-Homoproline hydrochloride, 98%
CAS: 53912-85-9 Molecular Formula: C6H12ClNO2 Molecular Weight (g/mol): 165.617 MDL Number: MFCD07363484 InChI Key: VQDACVOAOJQTPR-JEDNCBNOSA-N Synonym: s-2-pyrrolidin-2-yl acetic acid hydrochloride,l-beta-homoproline hydrochloride,l-beta-homoproline hcl,2-2s-pyrrolidin-2-yl acetic acid hydrochloride,2-pyrrolidineacetic acid, hydrochloride, 2s,s-2-2-pyrrolidinyl acetic acid hydrochloride,s-3-pyrrolidinecarboxylic acid hydrochloride,2s-pyrrolidin-2-ylacetic acid hydrochloride,l-beta-homoproline-hcl,s-3-pyrrol-idinecarboxylic acid hydrochloride PubChem CID: 2761540 IUPAC Name: 2-[(2S)-pyrrolidin-2-yl]acetic acid;hydrochloride SMILES: C1CC(NC1)CC(=O)O.Cl