accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 5392-12-1

IUPAC Name: 2-methoxynaphthalene-1-carbaldehyde
Synonyms: 2-methoxynaphthalene-1-carbaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: YIQGLTKAOHRZOL-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=CC2=C1C=O
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)