accessibility menu, dialog, popup

UserName

CAS RN 5393-99-7

SNN

CAS RN 5393-99-7

IUPAC Name: diphenyl-1,2,3-thiadiazole
Synonyms: diphenyl-1,2,3-thiadiazole
Molecular Weight (g/mol): 238.31
Molecular Formula: C14H10N2S
InChi Key: QVGVWLHVMVQIQI-UHFFFAOYSA-N
SMILES: S1N=NC(=C1C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results