accessibility menu, dialog, popup

UserName

CAS RN 5400-88-4

IUPAC Name: 4-tert-butylcyclohexan-1-amine
Synonyms: 4-tert-butylcyclohexan-1-amine
Molecular Weight (g/mol): 155.29
Molecular Formula: C10H21N
InChi Key: BGNLXETYTAAURD-UHFFFAOYSA-N
SMILES: CC(C)(C)C1CCC(N)CC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results