accessibility menu, dialog, popup

UserName

CH3H3CNH2O

CAS RN 541-46-8

IUPAC Name: 3-methylbutanamide
Synonyms: 3-methylbutanamide
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: SANOUVWGPVYVAV-UHFFFAOYSA-N
SMILES: CC(C)CC(N)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results