accessibility menu, dialog, popup

UserName

NH2SNHNH2S

CAS RN 541-53-7

IUPAC Name: (carbamothioylamino)methanethioamide
Synonyms: thioimidodicarbonic diamide
Molecular Weight (g/mol): 135.2
Molecular Formula: C2H5N3S2
InChi Key: JIRRNZWTWJGJCT-UHFFFAOYSA-N
SMILES: NC(=S)NC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results