accessibility menu, dialog, popup

UserName

HHClClNH2H2N

CAS RN 541-69-5

IUPAC Name: dihydrogen benzene-1,3-diamine dichloride
Synonyms: m-phenylenediamine hcl
Molecular Weight (g/mol): 181.06
Molecular Formula: C6H10Cl2N2
InChi Key: SVTOYMIYCMHPIV-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC(N)=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results