accessibility menu, dialog, popup

UserName

CAS RN 5413-75-2

NaNaOSOOONNHNNSOOO(Z)(E/Z)

CAS RN 5413-75-2

IUPAC Name: disodium (8Z)-7-oxo-8-{2-[4-(2-phenyldiazen-1-yl)phenyl]hydrazin-1-ylidene}-7,8-dihydronaphthalene-1,3-disulfonate
Synonyms: disodium (8Z)-7-oxo-8-{2-[4-(2-phenyldiazen-1-yl)phenyl]hydrazin-1-ylidene}naphthalene-1,3-disulfonate
Molecular Weight (g/mol): 556.47
Molecular Formula: C22H14N4Na2O7S2
InChi Key: OJPSNKJNYKPDPH-HKWNQDJZSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=C(C=C3)N=NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results