accessibility menu, dialog, popup

UserName

HOOOHN

CAS RN 5414-99-3

IUPAC Name: 4-(phenylcarbamoyl)butanoic acid
Synonyms: 4-(phenylcarbamoyl)butanoic acid
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: NUDGFGBJFZEEEJ-UHFFFAOYSA-N
SMILES: OC(=O)CCCC(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results