CAS RN 54261-98-2
CAS RN 54261-98-2
IUPAC Name:
2-({6-[(6-{[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl)methoxy]-3,4,5-trihydroxyoxan-2-yl}methoxy)-6-(hydroxymethyl)oxane-3,4,5-triol trihydrate
Synonyms:
2-({6-[(6-{[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl)methoxy]-3,4,5-trihydroxyoxan-2-yl}methoxy)-6-(hydroxymethyl)oxane-3,4,5-triol trihydrate
Molecular Weight (g/mol):
720.62
Molecular Formula:
C24H48O24
InChi Key:
XQDOUZBIGFWLCL-UHFFFAOYNA-N
SMILES:
O.O.O.OCC1OC(CO)(OC2OC(COC3OC(COC4OC(CO)C(O)C(O)C4O)C(O)C(O)C3O)C(O)C(O)C2O)C(O)C1O
1
–
2
of
2
results
1
Thermo Scientific Chemicals Stachyose hydrate, 97+%
CAS: 54261-98-2 Molecular Formula: C24H42O21 Molecular Weight (g/mol): 666.58 MDL Number: MFCD00149457 InChI Key: UQZIYBXSHAGNOE-CYDMKDQUNA-N PubChem CID: 131801001 SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
2
Stachyose hydrate, technical
CAS: 54261-98-2 Molecular Formula: C24H42O21 Molecular Weight (g/mol): 666.58 MDL Number: MFCD00149457 InChI Key: UQZIYBXSHAGNOE-CYDMKDQUNA-N PubChem CID: 131801001 SMILES: OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO[C@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O