accessibility menu, dialog, popup

UserName

CAS RN 54288-70-9

IUPAC Name: 4-bromopiperidin-1-ium
Synonyms: 4-bromopiperidin-1-ium
Molecular Weight (g/mol): 165.05
Molecular Formula: C5H11BrN
InChi Key: FIHYVUSUEHIGOM-UHFFFAOYSA-O
SMILES: BrC1CC[NH2+]CC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)