CAS RN 5435-36-9
CAS RN 5435-36-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Methylindole-3-acetic Acid, TRC
CAS: 5435-36-9 Molecular Formula: C11H11NO2 Molecular Weight (g/mol): 189.21 IUPAC Name: 2-(7-methyl-1H-indol-3-yl)acetic acid SMILES: Cc1cccc2c(CC(=O)O)c[nH]c12