CAS RN 54405-27-5
CAS RN 54405-27-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
MedChemExpress Mini Gastrin I, human
MedChemExpress Mini Gastrin I, human is a shorter version of human gastrin, consists of amino acids 5-17 of the parent peptide.
2
MedChemExpress Mini Gastrin I, human
MedChemExpress Mini Gastrin I, human is a shorter version of human gastrin, consists of amino acids 5-17 of the parent peptide.
3
MedChemExpress Mini Gastrin I, human
MedChemExpress Mini Gastrin I, human is a shorter version of human gastrin, consists of amino acids 5-17 of the parent peptide.