accessibility menu, dialog, popup

UserName

NONH2

CAS RN 54408-35-4

IUPAC Name: 1-(3-phenyl-1,2-oxazol-5-yl)methanamine
Synonyms: 1-(3-phenyl-1,2-oxazol-5-yl)methanamine
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: AQZLTCXQTOKUAA-UHFFFAOYSA-N
SMILES: NCC1=CC(=NO1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results