accessibility menu, dialog, popup

UserName

CAS RN 5445-26-1

IUPAC Name: ethyl 2-(4-nitrophenyl)acetate
Synonyms: ethyl 2-(4-nitrophenyl)acetate
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: DWDRNKYLWMKWTH-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results