accessibility menu, dialog, popup

UserName

OOOOOOOOOCeCe

CAS RN 54451-25-1

IUPAC Name: dicerium(3+) hydrate tricarbonate
Synonyms: dicerium(3+) hydrate tricarbonate
Molecular Weight (g/mol): 478.27
Molecular Formula: C3H2Ce2O10
InChi Key: KHSBAWXKALEJFR-UHFFFAOYSA-H
SMILES: O.[Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Physical Form 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)