accessibility menu, dialog, popup

UserName

ONOO

CAS RN 5455-98-1

IUPAC Name: 2-[(oxiran-2-yl)methyl]-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-(2,3-epoxypropyl)phthalimide
Molecular Weight (g/mol): 203.2
Molecular Formula: C11H9NO3
InChi Key: DUILGEYLVHGSEE-UHFFFAOYNA-N
SMILES: O=C1N(CC2CO2)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results