accessibility menu, dialog, popup

UserName

CAS RN 54678-23-8

IUPAC Name: λ¹-copper(1+) (methylsulfanyl)methane bromide
Synonyms: λ¹-copper(1+) dimethyl sulfide bromide
Molecular Weight (g/mol): 205.58
Molecular Formula: C2H6BrCuS
InChi Key: PMHQVHHXPFUNSP-UHFFFAOYSA-M
SMILES: [Cu+].[Br-].CSC
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results