accessibility menu, dialog, popup

UserName

CAS RN 54957-84-5

CH3NOH2NCl

CAS RN 54957-84-5

IUPAC Name: 2-chloro-6-methylpyridine-3-carboxamide
Synonyms: 2-chloro-6-methylpyridine-3-carboxamide
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: QFRWKKWOCPTEKS-UHFFFAOYSA-N
SMILES: CC1=NC(Cl)=C(C=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results