accessibility menu, dialog, popup

UserName

CAS RN 5504-65-4

IUPAC Name: 5-phenyl-1H-pyrazole-4-carboxylic acid
Synonyms: 3-phenyl-2H-pyrazole-4-carboxylic acid
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: LGTJKUYVFSBOMC-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(NN=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results