accessibility menu, dialog, popup

UserName

CAS RN 551-06-4

IUPAC Name: 1-isothiocyanatonaphthalene
Synonyms: ANIT kesscocide 1-naftylisothiokyanat ANI naphthalene, isothiocyanato- α-naphthysothiocyanate 1-isothiocyanonaphthalene isothiocyanato-naphthalene
Molecular Weight (g/mol): 185.24
Molecular Formula: C11H7NS
InChi Key: JBDOSUUXMYMWQH-UHFFFAOYSA-N
SMILES: S=C=NC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results