accessibility menu, dialog, popup

UserName

LiOHOO

CAS RN 552-38-5

IUPAC Name: lithium(1+) 2-hydroxybenzoate
Synonyms: lithium(1+) salicylate
Molecular Weight (g/mol): 144.05
Molecular Formula: C7H5LiO3
InChi Key: PSBOOKLOXQFNPZ-UHFFFAOYSA-M
SMILES: [Li+].OC1=CC=CC=C1C([O-])=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results