CAS RN 5520-66-1
IUPAC Name:
1-[4-(diethylamino)phenyl]ethan-1-one
Synonyms:
1-[4-(diethylamino)phenyl]ethanone
Molecular Weight (g/mol):
191.27
Molecular Formula:
C12H17NO
InChi Key:
HMIBQFXWSUBFTG-UHFFFAOYSA-N
SMILES:
CCN(CC)C1=CC=C(C=C1)C(C)=O
1
–
1
of
1
results
1
4'-Diethylaminoacetophenone, 99%
CAS: 5520-66-1 Molecular Formula: C12H17NO Molecular Weight (g/mol): 191.27 MDL Number: MFCD00043651 InChI Key: HMIBQFXWSUBFTG-UHFFFAOYSA-N Synonym: 4'-diethylaminoacetophenone,p-diethylaminoacetophenone,4-diethylaminoacetophenone,1-4-diethylamino phenyl ethanone,ethanone, 1-4-diethylamino phenyl,1-acetyl-4-diethylamino benzene,acmc-1aspz,4-acetyl-n,n-diethylaniline PubChem CID: 138529 IUPAC Name: 1-[4-(diethylamino)phenyl]ethanone SMILES: CCN(CC)C1=CC=C(C=C1)C(C)=O