accessibility menu, dialog, popup

UserName

CAS RN 5521-55-1

IUPAC Name: 5-methylpyrazine-2-carboxylic acid
Synonyms: 5-methylpyrazine-2-carboxylic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: RBYJWCRKFLGNDB-UHFFFAOYSA-N
SMILES: CC1=CN=C(C=N1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)