accessibility menu, dialog, popup

UserName

LiOO

CAS RN 553-54-8

IUPAC Name: lithium(1+) benzoate
Synonyms: lithium benzoate
Molecular Weight (g/mol): 128.06
Molecular Formula: C7H5LiO2
InChi Key: LDJNSLOKTFFLSL-UHFFFAOYSA-M
SMILES: [Li+].[O-]C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)