accessibility menu, dialog, popup

UserName

CH3S

CAS RN 554-14-3

IUPAC Name: 2-methylthiophene
Synonyms: 2-methylthiophene
Molecular Weight (g/mol): 98.16
Molecular Formula: C5H6S
InChi Key: XQQBUAPQHNYYRS-UHFFFAOYSA-N
SMILES: CC1=CC=CS1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results